2-[(1-Imidazolyl)methyl]aniline
Catalog # APOSOR470962-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:173.214 g/mol
Storage Temperature:Ambient
CAS Number:61292-50-0
Formula:C₁₀H₁₁N₃
MDL Number:MFCD09814189