Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ethyl 3-(4-Chloro-2-nitrophenyl)propanoate
Ethyl 3-(4-Chloro-2-nitrophenyl)propanoate
Ethyl 3-(4-Chloro-2-nitrophenyl)propanoate
APOSOR470931-1G
:
Apollo Scientific
:
Ethyl 3-(4-Chloro-2-nitrophenyl)propanoate
APOSOR470931-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
257.67 g/mol
:
Ambient
:
1185180-58-8
:
C₁₁H₁₂ClNO₄
:
MFCD27986822