N-Cyclopentyl-1-imidazolecarboxamide
Catalog # APOSOR470817-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:179.219 g/mol
Storage Temperature:Ambient
CAS Number:154696-60-3
Formula:C₉H₁₃N₃O
MDL Number:MFCD11099393