Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(2,4-Dichlorobenzyl)hydrazine dihydrochloride
(2,4-Dichlorobenzyl)hydrazine dihydrochloride
(2,4-Dichlorobenzyl)hydrazine dihydrochloride
APOSOR470716-5G
:
Apollo Scientific
:
(2,4-Dichlorobenzyl)hydrazine dihydrochloride
APOSOR470716-5G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 g
Seq:
3173
:
:
263.98 g/mol
:
Ambient
:
1469998-19-3
:
C₇H₈Cl₂N₂·2ClH
:
MFCD26937552