Methyl Morpholine-4-carboxylate
Catalog # APOSOR470715-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:145.156 g/mol
Storage Temperature:Ambient
CAS Number:6906-13-4
Formula:C₆H₁₁NO₃
MDL Number:MFCD00595228