Methyl (S)-2-(4-Hydroxyisoxazolidine-2-carbonyl)benzoate
Catalog # APOSOR470687-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:251.235 g/mol
Storage Temperature:Ambient
CAS Number:272459-61-7
Formula:C₁₂H₁₃NO₅
MDL Number:MFCD26523288