4-(2-Pyridyl)-3-butyn-1-ol
Catalog # APOSOR470620-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:147.174 g/mol
Storage Temperature:Ambient
CAS Number:395652-44-5
Formula:C₉H₉NO
MDL Number:MFCD18384524