3-(4-Pyridyl)-2-propen-1-ol
Catalog # APOSOR470566-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:135.163 g/mol
Storage Temperature:Ambient
CAS Number:193002-34-5
Formula:C₈H₉NO
MDL Number:MFCD18384354