Methyl 1-(2-Chlorophenyl)-4-oxocyclohexanecarboxylate
Catalog # APOSOR470533-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:266.72 g/mol
Storage Temperature:Ambient
CAS Number:1385694-76-7
Formula:C₁₄H₁₅ClO₃
MDL Number:MFCD22423139