Bestelling invoeren
Netherlands
ContactUsLinkComponent
5-[(3-Methoxyphenylamino)methylene]-2,2-dimethyl-1,3-dioxane-4,6-dione
5-[(3-Methoxyphenylamino)methylene]-2,2-dimethyl-1,3-dioxane-4,6-dione
Catalog # APOSOR470421-25G
Supplier:  Apollo Scientific
CAS Number:  
5-[(3-Methoxyphenylamino)methylene]-2,2-dimethyl-1,3-dioxane-4,6-dione
Catalog # APOSOR470421-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :277.273 g/mol
Storage Temperature:Ambient
CAS Number:213699-52-6
Formula:C₁₄H₁₅NO₅
MDL Number:MFCD02936591
Storage Temperature:Kamertemperatuur