5-(3-Methylphenyl)thiophene-2-carbaldehyde
Catalog # APOSOR470389-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:202.272 g/mol
Storage Temperature:Ambient
CAS Number:38401-69-3
Formula:C₁₂H₁₀OS
MDL Number:MFCD06410188