Order Entry
Netherlands
ContactUsLinkComponent
2-Amino-2-[6-(4-chlorophenyl)-3-pyridyl]acetic acid hydrochloride
2-Amino-2-[6-(4-chlorophenyl)-3-pyridyl]acetic acid hydrochloride
Catalog # APOSOR470345-5G
Supplier:  Apollo Scientific
CAS Number:  
2-Amino-2-[6-(4-chlorophenyl)-3-pyridyl]acetic acid hydrochloride
Catalog # APOSOR470345-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :299.153 g/mol
Storage Temperature:Ambient
CAS Number:1137666-85-3
Formula:C₁₃H₁₁ClN₂O₂·ClH
MDL Number:MFCD07371467