5-(2-Chlorophenyl)-3-pyridinecarbaldehyde
Catalog # APOSOR470281-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:217.651 g/mol
Storage Temperature:Ambient
CAS Number:855301-00-7
Formula:C₁₂H₈ClNO
MDL Number:MFCD06410222