Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ethyl 1,4-dioxaspiro[4.5]dec-8-ylideneacetate
Ethyl 1,4-dioxaspiro[4.5]dec-8-ylideneacetate 95+%
Ethyl 1,4-dioxaspiro[4.5]dec-8-ylideneacetate 95+%
Ethyl 1,4-dioxaspiro[4.5]dec-8-ylideneacetate 95+%
APOSOR46595-1G
:
Apollo Scientific
:
Ethyl 1,4-dioxaspiro[4.5]dec-8-ylideneacetate 95+%
APOSOR46595-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
226.269 g/mol
:
Ambient
:
51656-91-8
:
C₁₂H₁₈O₄
:
MFCD07779434