Order Entry
Netherlands
ContactUsLinkComponent
1H-Pyrazolo[3,4-b]pyridine-3-carboxaldehyde 97%
1H-Pyrazolo[3,4-b]pyridine-3-carboxaldehyde 97%
Catalog # APOSOR460010-1G
Supplier:  Apollo Scientific
CAS Number:  
1H-Pyrazolo[3,4-b]pyridine-3-carboxaldehyde 97%
Catalog # APOSOR460010-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :147.134 g/mol
Storage Temperature:Ambient
CAS Number:1010073-87-6
Formula:C₇H₅N₃O
MDL Number:MFCD11100578