
4-Chloro-1-methyl-1H-pyrazol-3-amine hydrochloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1197235-09-8
:
Formula:C₄H₆ClN₃·ClH
MDL Number:MFCD15142897
:168.024 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together