Order Entry
Netherlands
ContactUsLinkComponent
Methyl 5,6,7,8-tetrahydropyrazolo[5,1-b][1,3]oxazepine-2-carboxylate
Methyl 5,6,7,8-tetrahydropyrazolo[5,1-b][1,3]oxazepine-2-carboxylate
Catalog # APOSOR451246-1G
Supplier:  Apollo Scientific
CAS Number:  
Methyl 5,6,7,8-tetrahydropyrazolo[5,1-b][1,3]oxazepine-2-carboxylate
Catalog # APOSOR451246-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :196.203 g/mol
Storage Temperature:Ambient
CAS Number:1935360-89-6
Formula:C₉H₁₂N₂O₃
MDL Number:MFCD28975210