Order Entry
Netherlands
ContactUsLinkComponent
4-Boc-2-(bromomethyl)-morpholine
Irritant
4-Boc-2-(bromomethyl)-morpholine
Catalog # APOSOR451230-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4-Boc-2-(bromomethyl)-morpholine
Catalog # APOSOR451230-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :280.159 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:765914-78-1
Formula:C₁₀H₁₈BrNO₃
MDL Number:MFCD20489326