1-Boc-4-[(2S)-2-aminopropyl]-piperazine dihydrochloride
Catalog # APOSOR451098-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:316.268 g/mol
Storage Temperature:Ambient
CAS Number:1017606-58-4
Formula:C₁₂H₂₅N₃O₂·2ClH
MDL Number:MFCD28975218