(S)-2-(Methoxymethyl)-morpholine hydrochloride
Catalog # APOSOR451067-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:167.634 g/mol
Storage Temperature:Ambient
CAS Number:141196-38-5
Formula:C₆H₁₃NO₂·ClH
MDL Number:MFCD16294688