Order Entry
Netherlands
ContactUsLinkComponent
(S)-4-(2-Pyrrolidinylmethyl)morpholine dihydrochloride
(S)-4-(2-Pyrrolidinylmethyl)morpholine dihydrochloride
Catalog # APOSOR451053-1G
Supplier:  Apollo Scientific
CAS Number:  
(S)-4-(2-Pyrrolidinylmethyl)morpholine dihydrochloride
Catalog # APOSOR451053-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :243.174 g/mol
Storage Temperature:Ambient
CAS Number:91790-91-9
Formula:C₉H₁₈N₂O·2ClH
MDL Number:MFCD28975200