3-(4-Methoxyphenoxy)-azetidine hydrochloride
Catalog # APOSOR451034-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:215.677 g/mol
Storage Temperature:Ambient
CAS Number:1236862-34-2
Formula:C₁₀H₁₃NO₂·ClH
MDL Number:MFCD05664838