Order Entry
Netherlands
ContactUsLinkComponent
3-(4-Chlorophenoxy)azetidine hydrochloride
3-(4-Chlorophenoxy)azetidine hydrochloride
Catalog # APOSOR451023-1G
Supplier:  Apollo Scientific
CAS Number:  
3-(4-Chlorophenoxy)azetidine hydrochloride
Catalog # APOSOR451023-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :220.096 g/mol
Storage Temperature:Ambient
CAS Number:490021-97-1
Formula:C₉H₁₀ClNO·ClH
MDL Number:MFCD00277797