
3-(Hydroxymethyl)-4-methyl-1H-1,2,4-triazol-5(4H)-one 99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1182358-83-3
:
Formula:C₄H₇N₃O₂
MDL Number:MFCD22393449
:129.117 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together