Order Entry
Netherlands
ContactUsLinkComponent
5-{[2-(Methylamino)-5-nitrophenyl]amino}-5-oxopentanoic acid 97%
5-{[2-(Methylamino)-5-nitrophenyl]amino}-5-oxopentanoic acid 97%
Catalog # APOSOR40639-1G
Supplier:  Apollo Scientific
CAS Number:  
5-{[2-(Methylamino)-5-nitrophenyl]amino}-5-oxopentanoic acid 97%
Catalog # APOSOR40639-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :281.265 g/mol
Melting Pt:170.5…171.5 °C
Storage Temperature:Ambient
CAS Number:91644-13-2
Formula:C₁₂H₁₅N₃O₅
MDL Number:MFCD09840668