Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
tert-Butyl 4-bromobutanoate
tert-Butyl 4-bromobutanoate 96%
tert-Butyl 4-bromobutanoate 96%
APOSOR40573-25G
:
Apollo Scientific
:
tert-Butyl 4-bromobutanoate 96%
APOSOR40573-25G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
25 g
Seq:
3308
:
:
223.107 g/mol
:
GHS07
:
Ambient
:
110661-91-1
:
C₈H₁₅BrO₂
:
MFCD02179416