
4-(Bromomethyl)-3-iodobenzonitrile
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:182287-63-4
:
Formula:C₈H₅BrIN
MDL Number:MFCD16872046
ADR:8,III
:321.94 g/mol
UN:1759
:GHS05
Storage Temperature:Refrigerator
Frequently Bought Together