
3-Bromo-4-iodobenzoic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:249647-25-4
:
Formula:C₇H₄BrIO₂
MDL Number:MFCD06808825
:326.914 g/mol
Melting Pt:242…243 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together