Order Entry
Netherlands
ContactUsLinkComponent
N,2-Dimethylbutan-1-amine hydrochloride ≥98%
N,2-Dimethylbutan-1-amine hydrochloride ≥98%
Catalog # APOSOR361722-100MG
Supplier:  Apollo Scientific
CAS Number:  
N,2-Dimethylbutan-1-amine hydrochloride ≥98%
Catalog # APOSOR361722-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :137.651 g/mol
Storage Temperature:Ambient
CAS Number:130985-79-4
Formula:C₆H₁₅N·ClH
MDL Number:MFCD18483183