Order Entry
Netherlands
Orders LinkContactUsLinkComponent
3-(2-Methylphenyl)-1H-pyrazole

3-(2-Methylphenyl)-1H-pyrazole

Irritant
Supplier:   Apollo Scientific
3-(2-Methylphenyl)-1H-pyrazole
Supplier:  Apollo Scientific
Certificates
3-(2-Methylphenyl)-1H-pyrazole

2 Options Available Below

Product Details & Documents
Synonyms:  3-(o-Tolyl)-1H-pyrazole, 2-(1H-Pyrazol-3-yl)toluene
  • CAS Number:59843-49-1
  • Formula:C₁₀H₁₀N₂
  • MDL Number:MFCD03094644
  • Molecular Weight:158.2 g/mol
  • Melting Pt:46…48 °C
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    250 mg
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg - 65,20 €
    65,20 €
    1 * 250 mg
       65,20 €

  • Bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 189,00 €
    189,00 €
    1 * 1 g
       189,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.