Order Entry
Netherlands
ContactUsLinkComponent
3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1-tosyl-1H-pyrrolo[2,3-b]pyridine 97%
3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1-tosyl-1H-pyrrolo[2,3-b]pyridine 97%
Catalog # APOSOR350552-250MG
Supplier:  Apollo Scientific
CAS Number:  
3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1-tosyl-1H-pyrrolo[2,3-b]pyridine 97%
Catalog # APOSOR350552-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :398.284 g/mol
Storage Temperature:Ambient
CAS Number:866545-91-7
Formula:C₂₀H₂₃BN₂O₄S
MDL Number:MFCD11616456