(R)-2-Hydroxy-2-phenylacetohydrazide
Catalog # APOSOR350337-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:166.177 g/mol
Storage Temperature:Ambient
CAS Number:84049-61-6
Formula:C₈H₁₀N₂O₂
MDL Number:MFCD21332895