
Methyl 4-methyl-3-((4-(pyridin-3-yl)pyrimidin-2-yl)amino)benzoate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:917392-54-2
:
Formula:C₁₈H₁₆N₄O₂
MDL Number:MFCD16619370
:320.345 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together