
3-Methyl-isothiazole-5-sulphonic acid amide
Catalog # APOSOR346293-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:178.233 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:1022128-99-9
Formula:C₄H₆N₂O₂S₂
MDL Number:MFCD11036155