Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Pyridin-2-yl-thiophene-2-sulphonic acid amide
5-Pyridin-2-yl-thiophene-2-sulphonic acid amide
5-Pyridin-2-yl-thiophene-2-sulphonic acid amide
5-Pyridin-2-yl-thiophene-2-sulphonic acid amide
APOSOR346287-1G
:
Apollo Scientific
:
5-Pyridin-2-yl-thiophene-2-sulphonic acid amide
APOSOR346287-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
240.302 g/mol
:
Ambient
:
190659-63-3
:
C₉H₈N₂O₂S₂
:
MFCD00107905