6-tert-Butyl-nicotinonitrile
Catalog # APOSOR346171-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:160.216 g/mol
Storage Temperature:Ambient
CAS Number:56029-45-9
Formula:C₁₀H₁₂N₂
MDL Number:MFCD11845758