Order Entry
Netherlands
ContactUsLinkComponent
(R)-3,3'-Bis(4-nitrophenyl)-[1,1'-binaphthalene]-2,2'-diol
(R)-3,3'-Bis(4-nitrophenyl)-[1,1'-binaphthalene]-2,2'-diol
Catalog # APOSOR340012-250MG
Supplier:  Apollo Scientific
CAS Number:  
(R)-3,3'-Bis(4-nitrophenyl)-[1,1'-binaphthalene]-2,2'-diol
Catalog # APOSOR340012-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :528.511 g/mol
Storage Temperature:Freezer
CAS Number:791616-60-9
Formula:C₃₂H₂₀N₂O₆
MDL Number:MFCD28139323