3-Methyl-1-benzothiophen-2-amine hydrochloride, tech
Catalog # APOSOR33524-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:199.7 g/mol
Storage Temperature:Ambient
CAS Number:13584-56-0
Formula:C₉H₉NS·ClH
MDL Number:MFCD18904349