4-Methyl-3-(1-oxo-2,3-dihydro-1H-isoindol-2-yl)pentanoic acid, tech
Catalog # APOSOR33406-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:247.29 g/mol
Storage Temperature:Ambient
CAS Number:866144-51-6
Formula:C₁₄H₁₇NO₃
MDL Number:MFCD03305892