N'-[(Oxolan-3-yl)methyl](tert-butoxy)carbohydrazide, tech
Catalog # APOSOR33380-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:216.277 g/mol
Storage Temperature:Ambient
CAS Number:866143-73-9
Formula:C₁₀H₂₀N₂O₃
MDL Number:MFCD03305880