N-tert-Butyl-4-(6-chloropyridin-2-yl)piperazine-1-carboxamide, tech
Catalog # APOSOR33318-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:296.796 g/mol
Storage Temperature:Ambient
CAS Number:339107-28-7
Formula:C₁₄H₂₁ClN₄O
MDL Number:MFCD00139041