2-[(3-Chlorophenyl)methoxy]ethan-1-ol, tech
Catalog # APOSOR33269-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:186.635 g/mol
Storage Temperature:Ambient
CAS Number:478257-99-7
Formula:C₉H₁₁ClO₂
MDL Number:MFCD02187353