Order Entry
Netherlands
ContactUsLinkComponent
 
2-(Methylamino)-4-phenylpyridine-3-carbonitrile, tech
Catalog # APOSOR33249-1G
Supplier:  Apollo Scientific
CAS Number:  
2-(Methylamino)-4-phenylpyridine-3-carbonitrile, tech
Catalog # APOSOR33249-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :209.247 g/mol
Storage Temperature:Ambient
CAS Number:339103-54-7
Formula:C₁₃H₁₁N₃
MDL Number:MFCD01444225