Order Entry
Netherlands
ContactUsLinkComponent
(2E)-2-[2-(2,3-Dihydro-1,4-benzodioxin-6-yl)hydrazin-1-ylidene]cyclohexan-1-one, tech
(2E)-2-[2-(2,3-Dihydro-1,4-benzodioxin-6-yl)hydrazin-1-ylidene]cyclohexan-1-one, tech
Catalog # APOSOR33246-1G
Supplier:  Apollo Scientific
CAS Number:  
(2E)-2-[2-(2,3-Dihydro-1,4-benzodioxin-6-yl)hydrazin-1-ylidene]cyclohexan-1-one, tech
Catalog # APOSOR33246-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :260.288 g/mol
Storage Temperature:Ambient
CAS Number:339101-62-1
Formula:C₁₄H₁₆N₂O₃
MDL Number:MFCD01568696