Order Entry
Netherlands
ContactUsLinkComponent
 
1-(2,4-Dichlorophenyl)-2-methanesulphonylethan-1-one, tech
Catalog # APOSOR33235-1G
Supplier:  Apollo Scientific
CAS Number:  
1-(2,4-Dichlorophenyl)-2-methanesulphonylethan-1-one, tech
Catalog # APOSOR33235-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :267.129 g/mol
Storage Temperature:Ambient
CAS Number:186342-91-6
Formula:C₉H₈Cl₂O₃S
MDL Number:MFCD00202220