Order Entry
Netherlands
ContactUsLinkComponent
2,2,4-Trimethyl-2,3-dihydro-1H-1,5-benzodiazepine, tech
2,2,4-Trimethyl-2,3-dihydro-1H-1,5-benzodiazepine, tech
Catalog # APOSOR33171-1G
Supplier:  Apollo Scientific
CAS Number:  
2,2,4-Trimethyl-2,3-dihydro-1H-1,5-benzodiazepine, tech
Catalog # APOSOR33171-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :188.269 g/mol
Storage Temperature:Ambient
CAS Number:24107-34-4
Formula:C₁₂H₁₆N₂
MDL Number:MFCD00086184