9,9-Dimethyl-7,8,9,10-tetrahydrophenanthridin-7-one, tech
Catalog # APOSOR33170-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:225.286 g/mol
Storage Temperature:Ambient
CAS Number:70062-80-5
Formula:C₁₅H₁₅NO
MDL Number:MFCD03848688