Order Entry
Netherlands
ContactUsLinkComponent
1-(3-Methyl-4H-1,4-benzothiazin-2-yl)ethan-1-one 95%
1-(3-Methyl-4H-1,4-benzothiazin-2-yl)ethan-1-one 95%
Catalog # APOSOR33084-1G
Supplier:  Apollo Scientific
CAS Number:  
1-(3-Methyl-4H-1,4-benzothiazin-2-yl)ethan-1-one 95%
Catalog # APOSOR33084-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :205.276 g/mol
Storage Temperature:Ambient
CAS Number:31645-94-0
Formula:C₁₁H₁₁NOS
MDL Number:MFCD00053054