Methyl 6-(4-phenylpiperazin-1-yl)pyridine-3-carboxylate, tech
Catalog # APOSOR33078-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:297.352 g/mol
Storage Temperature:Ambient
CAS Number:132521-73-4
Formula:C₁₇H₁₉N₃O₂
MDL Number:MFCD01316590