Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Cyclopropanecarbonyl-2-cyclopropylpyridine
5-Cyclopropanecarbonyl-2-cyclopropylpyridine, tech
5-Cyclopropanecarbonyl-2-cyclopropylpyridine, tech
5-Cyclopropanecarbonyl-2-cyclopropylpyridine, tech
APOSOR33046-1G
:
Apollo Scientific
:
5-Cyclopropanecarbonyl-2-cyclopropylpyridine, tech
APOSOR33046-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
187.238 g/mol
:
Ambient
:
56260-49-2
:
C₁₂H₁₃NO
:
MFCD05256200